Assay Details
Target Gene Details
Entrez Gene ID: | 55233 |
Gene Name: | MOB kinase activator 1A |
Gene Aliases: |
C2orf6, MATS1, MOB1, MOBK1B, MOBKL1B, Mob4B |
Location: |
Chr.2:74152598-74178967 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MOB1A | NM_001317110.1 | 2 | 388 | NP_001304039.1 |
| NM_001317111.1 | 2 | 284 | NP_001304040.1 | |
| NM_001317112.1 | 2 | 391 | NP_001304041.1 | |
| NM_018221.4 | 2 | 391 | NP_060691.2 | |
| XM_011532948.2 | 2 | 251 | XP_011531250.1 | |
| AB016839.1 | 2 | 240 | BAB19058.1 | |
| AJ577474.1 | 1 | 98 | CAE12093.1 | |
| AK001650.1 | 2 | 281 | BAA91810.1 | |
| AK021657.1 | 2 | 305 | BAB13868.1 | |
| AK023321.1 | 2 | 251 | BAB14525.1 | |
| BC003398.1 | 2 | 292 | AAH03398.1 | |
| BI825399.1 | 2 | 284 | ||
| BU172616.1 | 2 | 287 | ||
| CN426611.1 | 2 | 279 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834261 | Chr.2:74164582 - 74386593 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map