Assay Details
Target Gene Details
Entrez Gene ID: | 85406 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member C14 |
Gene Aliases: |
DNAJ, DRIP78, HDJ3, LIP6 |
Location: |
Chr.12:55820960-55829636 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC14 | NM_032364.5 | 7 | 2263 | NP_115740.5 |
| AF141342.1 | 4 | 557 | AAG49445.1 | |
| AF351784.1 | 5 | 594 | AAK56241.1 | |
| AK055945.1 | 6 | 1301 | BAB71050.1 | |
| AK057354.1 | BAB71445.1 | |||
| AK123025.1 | 6 | 1843 | ||
| AK291222.1 | 7 | 2498 | ||
| AY188447.1 | 7 | 2180 | AAO73451.1 | |
| BC080655.1 | 6 | 1215 | AAH80655.1 | |
| BC117146.1 | 7 | 2134 | ||
| BC117148.1 | 7 | 2134 | ||
| EF560741.1 | 7 | 2185 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832423 | Chr.12:55687038 - 55859914 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map