Assay Details
Target Gene Details
Entrez Gene ID: | 64151 |
Gene Name: | non-SMC condensin I complex subunit G |
Gene Aliases: |
CAPG, CHCG, NY-MEL-3, YCG1 |
Location: |
Chr.4:17810813-17844865 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCAPG | NM_022346.4 | 1 | 150 | NP_071741.2 |
| NR_073124.1 | 1 | 150 | ||
| XM_011513876.2 | XP_011512178.1 | |||
| XM_017008543.1 | XP_016864032.1 | |||
| XM_017008544.1 | XP_016864033.1 | |||
| AB013299.1 | BAB21557.1 | |||
| AF235023.1 | AAG30732.1 | |||
| AF331796.1 | 1 | 61 | AAG49627.1 | |
| AK022512.1 | BAB14069.1 | |||
| BC068467.1 | AAH68467.1 | |||
| BC101476.1 | AAI01477.1 | |||
| BX474610.1 | ||||
| DB451361.1 | 1 | 150 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893889 | Chr.4:17807915 - 17817512 on Build GRCh38 | Loss |
|
| esv2645526 | Chr.4:17810033 - 17811527 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs117540091] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map