Assay Details
Target Gene Details
Entrez Gene ID: | 23787 |
Gene Name: | mitochondrial carrier 1 |
Gene Aliases: |
CGI-64, PIG60, PSAP, SLC25A49 |
Location: |
Chr.6:36968135-36986551 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTCH1 | NM_001271641.1 | NP_001258570.1 | ||
| NM_014341.2 | NP_055156.1 | |||
| NR_130739.1 | ||||
| XM_005248978.1 | XP_005249035.1 | |||
| XM_005248979.1 | XP_005249036.1 | |||
| XM_011514450.1 | XP_011512752.1 | |||
| XM_017010642.1 | XP_016866131.1 | |||
| XM_017010643.1 | XP_016866132.1 | |||
| AF151822.1 | AAD34059.1 | |||
| AF176006.3 | AAD52644.3 | |||
| AF189289.1 | AAF07936.1 | |||
| AF192559.3 | AAF12793.3 | |||
| AK022783.1 | ||||
| AK130842.1 | ||||
| AK225628.1 | ||||
| AK303318.1 | ||||
| AK315067.1 | ||||
| AY358519.1 | AAQ88883.1 | |||
| AY947344.1 | ||||
| BC000702.2 | AAH00702.2 | |||
| BC006507.2 | AAH06507.1 | |||
| BC009962.2 | AAH09962.2 | |||
| BC040659.1 | AAH40659.2 | |||
| BC110914.1 | AAI10915.1 | |||
| BC125056.1 | ||||
| BC153873.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1015689 | Chr.6:36550916 - 39052434 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map