Assay Details
Target Gene Details
Entrez Gene ID: | 284716 |
Gene Name: | ribosomal modification protein rimK like family member A |
Gene Aliases: |
FAM80A, NAAGS, NAAGS-II |
Location: |
Chr.1:42380797-42425813 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RIMKLA | NM_173642.3 | 5 | 908 | NP_775913.2 |
| XM_006710585.3 | 5 | 786 | XP_006710648.1 | |
| XM_017001082.1 | 6 | 967 | XP_016856571.1 | |
| BC039737.1 | 5 | 902 | AAH39737.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv945902 | Chr.1:42412295 - 42414555 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map