Assay Details
Target Gene Details
Entrez Gene ID: | 5431 |
Gene Name: | RNA polymerase II subunit B |
Gene Aliases: |
POL2RB, RPB2, hRPB140 |
Location: |
Chr.4:56978640-57031162 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLR2B | NM_000938.2 | 8 | 1294 | NP_000929.1 |
| NM_001303268.1 | 7 | 1143 | NP_001290197.1 | |
| NM_001303269.1 | 9 | 1472 | NP_001290198.1 | |
| AK289823.1 | 8 | 1038 | ||
| AK294901.1 | 7 | 919 | ||
| AK295132.1 | 9 | 1243 | ||
| AK299090.1 | 8 | 810 | ||
| BC023503.2 | 8 | 1052 | AAH23503.2 | |
| X63563.1 | 8 | 991 | CAA45124.1 |
More Information
Set Membership: |
|

Genomic Map