Assay Details
Target Gene Details
Entrez Gene ID: | 115752 |
Gene Name: | DIS3 like exosome 3'-5' exoribonuclease |
Gene Aliases: |
DIS3L1 |
Location: |
Chr.15:66293257-66333898 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DIS3L | NM_001323936.1 | NP_001310865.1 | ||
| NM_001323937.1 | 1 | 64 | NP_001310866.1 | |
| NM_133375.4 | 1 | 64 | NP_588616.1 | |
| AK295392.1 | ||||
| BC022089.2 | AAH22089.1 | |||
| BG722405.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1047176 | Chr.15:66036143 - 66310781 on Build GRCh38 | Gain |
|
| nsv1047588 | Chr.15:66227169 - 66394432 on Build GRCh38 | Loss |
|
| nsv569793 | Chr.15:66047062 - 66308291 on Build GRCh38 | Gain |
|
| nsv1055090 | Chr.15:66217529 - 66298669 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map