Assay Details
Target Gene Details
Entrez Gene ID: | 115584 |
Gene Name: | solute carrier family 5 member 11 |
Gene Aliases: |
KST1, RKST1, SGLT6, SMIT2 |
Location: |
Chr.16:24832784-24911711 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833175 | Chr.16:24846570 - 25080507 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map