Assay Details
Target Gene Details
Entrez Gene ID: | 440400 |
Gene Name: | ribonuclease K |
Gene Aliases: |
- |
Location: |
Chr.17:7012417-7014533 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNASEK | NM_001004333.4 | NP_001004333.2 | ||
| NR_037715.1 | ||||
| NR_037716.1 | ||||
| AA633512.1 | ||||
| AA843796.1 | ||||
| AK289930.1 | ||||
| BC051802.1 | AAH51802.1 | |||
| BC062705.1 | AAH62705.1 | |||
| BC095436.1 | AAH95436.1 | |||
| BQ223829.1 | ||||
| CD176288.1 |
Target Gene Details
Entrez Gene ID: | 100529209 |
Gene Name: | RNASEK-C17orf49 readthrough |
Gene Aliases: |
- |
Location: |
Chr.17:7012417-7017524 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNASEK-C17orf49 | NR_037717.1 | 3 | 776 | |
| BC040148.1 | 3 | 533 | AAH40148.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422288 | Chr.17:6789933 - 7646709 on Build GRCh38 | Deletion |
|
| nsv952117 | Chr.17:7014482 - 7028381 on Build GRCh38 | Deletion |
|
| nsv457659 | Chr.17:7010333 - 7689462 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs117114327,rs557159990] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map