Assay Details
Target Gene Details
Entrez Gene ID: | 121643 |
Gene Name: | forkhead box N4 |
Gene Aliases: |
- |
Location: |
Chr.12:109277978-109309406 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FOXN4 | NM_213596.2 | 9 | 1113 | NP_998761.2 |
| XM_011537922.2 | 9 | 1302 | XP_011536224.1 | |
| XM_017018818.1 | 7 | 2088 | XP_016874307.1 | |
| AF425596.1 | 7 | 888 | AAL23949.1 | |
| AF425597.1 | 6 | 763 | AAL23950.1 | |
| AK093286.1 | ||||
| AK131519.1 | 5 | 720 | BAD18661.1 | |
| BC113073.1 | 2 | 113 | ||
| BC146825.1 | 9 | 1114 |
More Information
Set Membership: |
|

Genomic Map