Assay Details
Target Gene Details
Entrez Gene ID: | 23621 |
Gene Name: | beta-secretase 1 |
Gene Aliases: |
ASP2, BACE, HSPC104 |
Location: |
Chr.11:117285686-117316256 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BACE1 | NM_001207048.1 | 8 | 4704 | NP_001193977.1 |
| NM_001207049.1 | 8 | 4629 | NP_001193978.1 | |
| NM_012104.4 | 9 | 5342 | NP_036236.1 | |
| NM_138971.3 | 9 | 5210 | NP_620427.1 | |
| NM_138972.3 | 9 | 5267 | NP_620428.1 | |
| NM_138973.3 | 9 | 5135 | NP_620429.1 | |
| AB032975.2 | 9 | 5324 | BAA86463.2 | |
| AB177847.1 | 8 | 4566 | BAD66825.1 | |
| AF201468.1 | 9 | 5335 | AAF18982.1 | |
| BM996673.1 | 1 | 523 | ||
| BQ005204.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509443 | Chr.11:117255121 - 117293441 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map