Assay Details
Target Gene Details
Entrez Gene ID: | 23659 |
Gene Name: | phospholipase A2 group XV |
Gene Aliases: |
ACS, GXVPLA2, LLPL, LPLA2, LYPLA3 |
Location: |
Chr.16:68245344-68261062 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLA2G15 | NM_012320.3 | 6 | 1662 | NP_036452.1 |
| XM_005255886.3 | 6 | 1510 | XP_005255943.1 | |
| XM_011522979.2 | 7 | 1731 | XP_011521281.1 | |
| XM_011522980.2 | 7 | 1602 | XP_011521282.2 | |
| AK001705.1 | 6 | 1623 | ||
| AK074828.1 | 6 | 1662 | BAC11233.1 | |
| AK127898.1 | 2 | 2062 | ||
| AL110209.1 | 6 | 1643 | CAB53675.1 | |
| AL389957.1 | 2 | 919 | CAB97531.1 | |
| AY358425.1 | 6 | 1599 | AAQ88791.1 | |
| BC011640.2 | 3 | 1176 | AAH11640.2 | |
| BC062605.1 | 6 | 1626 | AAH62605.1 |
More Information
Set Membership: |
|

Genomic Map