Assay Details
Target Gene Details
Entrez Gene ID: | 29937 |
Gene Name: | neudesin neurotrophic factor |
Gene Aliases: |
CIR2, SCIRP10, SPUF |
Location: |
Chr.1:212432887-212446379 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NENF | NM_013349.4 | 4 | 613 | NP_037481.1 |
| NR_026598.1 | 3 | 552 | ||
| AB126219.1 | BAD72063.1 | |||
| AF000267.1 | 1 | 118 | ||
| AF049672.1 | 4 | 411 | ||
| AF173937.1 | 4 | 616 | AAD51419.1 | |
| AK074431.1 | 4 | 581 | ||
| AK223135.1 | 4 | 581 | BAD96855.1 | |
| AY762102.1 | 4 | 572 | ||
| BC008823.2 | 4 | 562 | AAH08823.1 | |
| DB502983.1 | ||||
| DB574171.1 | 1 | 123 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv549139 | Chr.1:212158952 - 212703560 on Build GRCh38 | Gain |
|
| nsv527863 | Chr.1:212444081 - 212527276 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map