Assay Details
Target Gene Details
Entrez Gene ID: | 54555 |
Gene Name: | DEAD-box helicase 49 |
Gene Aliases: |
R27090_2 |
Location: |
Chr.19:18919675-18928633 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX49 | NM_019070.4 | 6 | 803 | NP_061943.2 |
| NR_033677.1 | 6 | 732 | ||
| XM_011528083.1 | 7 | 851 | XP_011526385.1 | |
| XM_011528084.2 | 7 | 795 | XP_011526386.1 | |
| AK001294.1 | 6 | 752 | BAA91606.1 | |
| AK223294.1 | 6 | 746 | BAD97014.1 | |
| AK291916.1 | 6 | 763 | ||
| AL110272.1 | 2 | 869 | CAH10729.1 | |
| AL137702.1 | 2 | 869 | CAH10707.1 | |
| BC000979.3 | 7 | 827 | AAH00979.2 | |
| BC002674.2 | 6 | 793 | AAH02674.1 | |
| BX647408.1 | 7 | 670 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470132 | Chr.19:18853633 - 18943911 on Build GRCh38 | Loss |
|
| nsv833778 | Chr.19:18901860 - 19090678 on Build GRCh38 | Loss |
|
| nsv1151217 | Chr.19:18627369 - 19284914 on Build GRCh38 | Deletion |
|
| nsv952423 | Chr.19:18859292 - 18941091 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map