Assay Details
Target Gene Details
Entrez Gene ID: | 4978 |
Gene Name: | opioid binding protein/cell adhesion molecule like |
Gene Aliases: |
IGLON1, OBCAM, OPCM |
Location: |
Chr.11:132403361-133532983 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OPCML | NM_001012393.2 | 8 | 4426 | NP_001012393.1 |
| NM_001319103.1 | 8 | 4731 | NP_001306032.1 | |
| NM_001319104.1 | 7 | 4341 | NP_001306033.1 | |
| NM_001319105.1 | 8 | 4250 | NP_001306034.1 | |
| NM_001319106.1 | 7 | 4701 | NP_001306035.1 | |
| NM_002545.4 | 7 | 4704 | NP_002536.1 | |
| XM_006718846.2 | 8 | 4649 | XP_006718909.1 | |
| XM_011542856.2 | 9 | 4962 | XP_011541158.1 | |
| AK125185.1 | 1 | 967 | ||
| BX537377.1 | 8 | 4274 | CAD97619.1 |
More Information
Set Membership: |
|

Genomic Map