Assay Details
Target Gene Details
Entrez Gene ID: | 124935 |
Gene Name: | solute carrier family 43 member 2 |
Gene Aliases: |
LAT4 |
Location: |
Chr.17:1569254-1630014 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC43A2 | NM_001284498.1 | 15 | 2823 | NP_001271427.1 |
| NM_001284499.1 | 10 | 2264 | NP_001271428.1 | |
| NM_001321364.1 | 15 | 2621 | NP_001308293.1 | |
| NM_001321365.1 | 15 | 2790 | NP_001308294.1 | |
| NM_152346.2 | 14 | 2609 | NP_689559.1 | |
| XM_017024177.1 | 14 | 2786 | XP_016879666.1 | |
| XM_017024178.1 | 14 | 2819 | XP_016879667.1 | |
| XM_017024179.1 | 13 | 2718 | XP_016879668.1 | |
| AB120364.1 | 15 | 2789 | BAD91091.1 | |
| AK074428.1 | 4 | 1140 | ||
| AK075068.1 | 12 | 2149 | BAC11383.1 | |
| AK075173.1 | 12 | 2203 | BAC11450.1 | |
| AK075196.1 | 8 | 1890 | BAC11464.1 | |
| BC019562.1 | 8 | 1884 | AAH19562.1 | |
| BC027923.1 | 14 | 2559 | AAH27923.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833334 | Chr.17:1411327 - 1582381 on Build GRCh38 | Loss |
|
| nsv1945 | Chr.17:1560608 - 1589052 on Build GRCh38 | Insertion |
|
| esv1004030 | Chr.17:1566596 - 1574887 on Build GRCh38 | Deletion |
|
| nsv457635 | Chr.17:1554545 - 1638896 on Build GRCh38 | Gain |
|
| esv2670177 | Chr.17:1572448 - 1587683 on Build GRCh38 | Deletion |
|
| esv2715472 | Chr.17:1572813 - 1588114 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map