Assay Details
Target Gene Details
Entrez Gene ID: | 7159 |
Gene Name: | tumor protein p53 binding protein 2 |
Gene Aliases: |
53BP2, ASPP2, BBP, P53BP2, PPP1R13A |
Location: |
Chr.1:223779893-223845981 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TP53BP2 | NM_001031685.2 | 17 | 3503 | NP_001026855.2 |
| NM_005426.2 | 18 | 3635 | NP_005417.1 | |
| XM_011544268.1 | 16 | 3364 | XP_011542570.1 | |
| XM_011544269.2 | 14 | 3022 | XP_011542571.1 | |
| XM_017002220.1 | 18 | 3644 | XP_016857709.1 | |
| AJ318888.2 | 17 | 3448 | CAC83012.1 | |
| AK294432.1 | 17 | 3355 | ||
| AK294654.1 | 7 | 1439 | ||
| AK294964.1 | 10 | 2523 | ||
| AK295378.1 | 11 | 2227 | ||
| AK316016.1 | 10 | 2425 | ||
| AK316103.1 | 10 | 2403 | ||
| BC040247.1 | 15 | 2979 | ||
| BC058918.1 | 17 | 3461 | ||
| U09582.1 | 6 | 1398 | AAA21597.1 | |
| U58334.1 | 18 | 3580 | AAC50557.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4698 | Chr.1:223770507 - 223787519 on Build GRCh38 | Insertion |
|
| esv34082 | Chr.1:223663316 - 223956807 on Build GRCh38 | Loss |
|
| esv2759000 | Chr.1:223747518 - 224175461 on Build GRCh38 | Gain+Loss |
|
| nsv832681 | Chr.1:223524706 - 223814989 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map