Assay Details
Target Gene Details
Entrez Gene ID: | 203102 |
Gene Name: | ADAM metallopeptidase domain 32 |
Gene Aliases: |
- |
Location: |
Chr.8:39107529-39284917 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAM32 | NM_001313994.1 | 1 | 216 | NP_001300923.1 |
| NM_145004.6 | 1 | 216 | NP_659441.3 | |
| XM_006716298.2 | XP_006716361.1 | |||
| XM_006716299.1 | XP_006716362.1 | |||
| XM_011544433.1 | XP_011542735.1 | |||
| XM_011544434.1 | XP_011542736.1 | |||
| XM_017013183.1 | XP_016868672.1 | |||
| XM_017013184.1 | XP_016868673.1 | |||
| XM_017013186.1 | XP_016868675.1 | |||
| AK122608.1 | BAC85489.1 | |||
| AK300148.1 | 1 | 174 | ||
| AK313864.1 | 1 | 37 | ||
| AY172952.1 | 1 | 138 | AAO45696.1 | |
| AY358739.1 | 1 | 96 | AAQ89099.1 | |
| BC026085.2 | AAH26085.1 | |||
| BC026169.2 | AAH26169.1 | |||
| BC028702.2 | AAH28702.1 | |||
| BC030014.2 | AAH30014.1 | |||
| BC030698.2 | AAH30698.1 | |||
| BC034975.2 | AAH34975.1 | |||
| BI559590.1 | 1 | 221 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6162 | Chr.8:39101789 - 39134363 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map