Assay Details
Target Gene Details
Entrez Gene ID: | 14 |
Gene Name: | angio associated migratory cell protein |
Gene Aliases: |
- |
Location: |
Chr.2:218264129-218270209 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AAMP | NM_001087.4 | NP_001078.2 | ||
| NM_001302545.1 | NP_001289474.1 | |||
| AB209790.1 | BAD93027.1 | |||
| AK131047.1 | ||||
| AK225078.1 | ||||
| BC020244.1 | AAH20244.1 | |||
| BC039866.1 | AAH39866.2 | |||
| BG534787.1 | ||||
| CN277005.1 | ||||
| CR456755.1 | CAG33036.1 | |||
| DA277259.1 | ||||
| DA465288.1 | ||||
| M95627.1 | AAA68889.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs74349235] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map