Assay Details
Target Gene Details
Entrez Gene ID: | 163590 |
Gene Name: | torsin 1A interacting protein 2 |
Gene Aliases: |
IFRG15, LULL1, NET9 |
Location: |
Chr.1:179839967-179878614 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TOR1AIP2 | NM_001199260.1 | 7 | 2233 | NP_001186189.1 |
| NM_145034.4 | 6 | 1814 | NP_659471.1 | |
| XM_005244939.4 | 7 | 2206 | XP_005244996.1 | |
| XM_017000474.1 | 8 | 2351 | XP_016855963.1 | |
| XM_017000475.1 | 6 | 2075 | XP_016855964.1 | |
| XM_017000476.1 | 3 | 1515 | XP_016855965.1 | |
| AF464140.1 | 6 | 1808 | AAM50514.1 | |
| BC101532.1 | 4 | 1518 | AAI01533.1 | |
| BC112225.1 | 4 | 1518 | AAI12226.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv473n100 | Chr.1:179814787 - 179895575 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map