Assay Details
Target Gene Details
Entrez Gene ID: | 987 |
Gene Name: | LPS responsive beige-like anchor protein |
Gene Aliases: |
BGL, CDC4L, CVID8, LAB300, LBA |
Location: |
Chr.4:150264515-151015725 on Build GRCh38 |
Assay Gene Location: | Within Intron 42 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRBA | NM_001199282.2 | NP_001186211.2 | ||
| NM_006726.4 | NP_006717.2 | |||
| XM_005263372.3 | XP_005263429.1 | |||
| XM_005263373.2 | XP_005263430.1 | |||
| XM_005263374.3 | XP_005263431.1 | |||
| XM_005263375.3 | XP_005263432.1 | |||
| XM_011532434.2 | XP_011530736.1 | |||
| XM_017008872.1 | XP_016864361.1 | |||
| AB208838.1 | BAD92075.1 | |||
| AF216648.2 | AAG48558.2 | |||
| AF467287.1 | AAM53530.1 | |||
| AK122920.1 | ||||
| M83822.1 | AAB09603.1 |
Target Gene Details
Entrez Gene ID: | 10586 |
Gene Name: | mab-21 like 2 |
Gene Aliases: |
MCOPS14 |
Location: |
Chr.4:150581925-150584693 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAB21L2 | NM_006439.4 | 1 | 380 | NP_006430.1 |
| AF262032.1 | AAG09800.1 | |||
| AK055665.1 | 1 | 154 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1024820 | Chr.4:150442926 - 151022409 on Build GRCh38 | Gain |
|
| nsv4547 | Chr.4:150565128 - 150610327 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map