Assay Details
Target Gene Details
Entrez Gene ID: | 23457 |
Gene Name: | ATP binding cassette subfamily B member 9 |
Gene Aliases: |
EST122234, TAPL |
Location: |
Chr.12:122918708-122975160 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCB9 | NM_001243013.1 | NP_001229942.1 | ||
| NM_001243014.1 | 8 | 1821 | NP_001229943.1 | |
| NM_019624.3 | 7 | 1692 | NP_062570.1 | |
| NM_019625.3 | 8 | 1821 | NP_062571.1 | |
| NM_203444.3 | 8 | 1821 | NP_982269.2 | |
| XM_005253558.2 | 8 | 1761 | XP_005253615.1 | |
| XM_011538095.2 | 8 | 2044 | XP_011536397.1 | |
| XM_011538096.2 | 8 | 1840 | XP_011536398.1 | |
| XM_011538098.2 | 7 | 2686 | XP_011536400.1 | |
| XM_011538099.2 | XP_011536401.1 | |||
| XM_017019103.1 | 9 | 1882 | XP_016874592.1 | |
| XM_017019104.1 | XP_016874593.1 | |||
| AB040953.2 | 8 | 4350 | ||
| AB045381.2 | 7 | 1570 | BAA97989.2 | |
| AB112582.1 | 8 | 1772 | BAC98409.1 | |
| AB112583.1 | 8 | 1772 | BAC98410.1 | |
| AB177852.1 | 8 | 1733 | ||
| AF216494.1 | 8 | 1809 | AAF89993.1 | |
| AK027624.1 | ||||
| AK074738.1 | 5 | 765 | BAC11171.1 | |
| AK294225.1 | 2 | 380 | ||
| AK304295.1 | ||||
| AL833963.1 | CAD38811.1 | |||
| BC017348.2 | 7 | 1536 | AAH17348.1 | |
| BC064384.1 | 8 | 2089 | AAH64384.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv22n68 | Chr.12:122911380 - 123104743 on Build GRCh38 | Loss |
|
| nsv517200 | Chr.12:122827504 - 123169951 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map