Assay Details
Target Gene Details
Entrez Gene ID: | 4209 |
Gene Name: | myocyte enhancer factor 2D |
Gene Aliases: |
- |
Location: |
Chr.1:156463721-156500842 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MEF2D | NM_001271629.1 | NP_001258558.1 | ||
| NM_005920.3 | NP_005911.1 | |||
| XM_005245169.4 | XP_005245226.1 | |||
| XM_005245170.3 | XP_005245227.1 | |||
| XM_006711330.3 | XP_006711393.1 | |||
| XM_006711332.3 | XP_006711395.1 | |||
| XM_006711333.2 | XP_006711396.1 | |||
| XM_006711334.3 | XP_006711397.1 | |||
| XM_011509569.2 | XP_011507871.1 | |||
| XM_017001314.1 | XP_016856803.1 | |||
| XM_017001315.1 | XP_016856804.1 | |||
| AB210005.1 | BAE06087.1 | |||
| AK308641.1 | ||||
| BC032479.1 | ||||
| BC040949.1 | AAH40949.1 | |||
| BC054520.1 | AAH54520.1 | |||
| L16794.1 | AAA93194.1 |
More Information
Set Membership: |
|

Genomic Map