Assay Details
Target Gene Details
Entrez Gene ID: | 57538 |
Gene Name: | alpha kinase 3 |
Gene Aliases: |
MAK, MIDORI |
Location: |
Chr.15:84816631-84873482 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALPK3 | NM_020778.4 | NP_065829.3 | ||
| XM_017022444.1 | XP_016877933.1 | |||
| XM_017022445.1 | XP_016877934.1 | |||
| AY044449.1 | AAK95951.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2760049 | Chr.15:84719698 - 84873299 on Build GRCh38 | Loss |
|
| esv2672879 | Chr.15:84183834 - 85188520 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map