Assay Details
Target Gene Details
Entrez Gene ID: | 26005 |
Gene Name: | C2 calcium dependent domain containing 3 |
Gene Aliases: |
OFD14 |
Location: |
Chr.11:74012578-74171210 on Build GRCh38 |
Assay Gene Location: | Within Exon 33 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C2CD3 | NM_001286577.1 | 30 | 6063 | NP_001273506.1 |
| NM_015531.5 | 30 | 6063 | NP_056346.3 | |
| XM_011544904.1 | 30 | 5962 | XP_011543206.1 | |
| XM_011544905.1 | 30 | 5962 | XP_011543207.1 | |
| XM_017017510.1 | 30 | 5962 | XP_016872999.1 | |
| XM_017017511.1 | 29 | 5888 | XP_016873000.1 | |
| XM_017017512.1 | 29 | 5853 | XP_016873001.1 | |
| XM_017017513.1 | 20 | 4059 | XP_016873002.1 | |
| XM_017017515.1 | 16 | 3612 | XP_016873004.1 | |
| XM_017017516.1 | 16 | 3182 | XP_016873005.1 | |
| AB231764.1 | 30 | 5877 | BAE17137.1 | |
| AI873280.1 | 2 | 227 | ||
| AK125894.1 | 17 | 3724 | BAC86334.1 | |
| AL833903.1 | 11 | 2260 | CAD38759.1 | |
| BC035599.1 | 10 | 2068 | AAH35599.1 | |
| BC110508.2 | 17 | 3692 | AAI10509.1 | |
| CD369754.1 | 2 | 238 |
More Information
Set Membership: |
|

Genomic Map