Assay Details
Target Gene Details
Entrez Gene ID: | 352909 |
Gene Name: | dynein axonemal assembly factor 3 |
Gene Aliases: |
C19orf51, CILD2, DAB1, PCD, PF22 |
Location: |
Chr.19:55158661-55166722 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAAF3 | NM_001256714.1 | NP_001243643.1 | ||
| NM_001256715.1 | NP_001243644.1 | |||
| NM_001256716.1 | NP_001243645.1 | |||
| NM_178837.4 | NP_849159.2 | |||
| AK093458.1 | BAC04172.1 | |||
| AK093473.1 | BAC04177.1 | |||
| AK097388.1 | BAC05029.1 | |||
| AW182162.1 | ||||
| BC016843.1 | AAH16843.1 | |||
| BC063449.1 | AAH63449.1 |
More Information
Set Membership: |
|

Genomic Map