Assay Details
Target Gene Details
Entrez Gene ID: | 55723 |
Gene Name: | anti-silencing function 1B histone chaperone |
Gene Aliases: |
CIA-II |
Location: |
Chr.19:14119509-14136628 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASF1B | NM_018154.2 | NP_060624.1 | ||
| AF279307.1 | AAK82973.1 | |||
| AK001288.1 | BAA91602.1 | |||
| AK001466.1 | BAA91708.1 | |||
| AK223080.1 | BAD96800.1 | |||
| AK302134.1 | ||||
| BC007726.2 | AAH07726.1 | |||
| BC010014.2 | AAH10014.1 | |||
| BC036521.1 | AAH36521.1 | |||
| CN426445.1 | 1 | 400 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833761 | Chr.19:13998659 - 14136778 on Build GRCh38 | Loss |
|
| nsv953980 | Chr.19:14082589 - 14210288 on Build GRCh38 | Deletion |
|
| nsv828452 | Chr.19:14059742 - 14148154 on Build GRCh38 | Loss |
|
| dgv49n68 | Chr.19:14066502 - 14274131 on Build GRCh38 | Loss |
|
| nsv517436 | Chr.19:14135617 - 14187630 on Build GRCh38 | Loss |
|
| nsv578642 | Chr.19:14028192 - 14187630 on Build GRCh38 | Gain |
|
| nsv578663 | Chr.19:14111697 - 14226588 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs555154075] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map