Assay Details
Target Gene Details
Entrez Gene ID: | 10682 |
Gene Name: | emopamil binding protein (sterol isomerase) |
Gene Aliases: |
CDPX2, CHO2, CPX, CPXD, MEND |
Location: |
Chr.X:48521776-48528716 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EBP | NM_006579.2 | NP_006570.1 | ||
| AU128761.1 | ||||
| BC001549.1 | AAH01549.1 | |||
| BC001572.1 | AAH01572.1 | |||
| BC046501.1 | AAH46501.1 | |||
| CA488777.1 | ||||
| CR456815.1 | CAG33096.1 | |||
| CR542094.1 | CAG46891.1 | |||
| KJ921656.1 | ||||
| Z37986.1 | CAA86068.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528292 | Chr.X:48433286 - 49274368 on Build GRCh38 | Gain |
|
| esv32909 | Chr.X:48003374 - 49285646 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map