Assay Details
Target Gene Details
Entrez Gene ID: | 4201 |
Gene Name: | male-enhanced antigen 1 |
Gene Aliases: |
HYS, MEA |
Location: |
Chr.6:42980344-43016886 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MEA1 | XM_017010868.1 | XP_016866357.1 | ||
| AK309759.1 |
Target Gene Details
Entrez Gene ID: | 5528 |
Gene Name: | protein phosphatase 2 regulatory subunit B'delta |
Gene Aliases: |
B56D, MRD35 |
Location: |
Chr.6:42984499-43012345 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP2R5D | NM_001270476.1 | 3 | 470 | NP_001257405.1 |
| NM_006245.3 | 3 | 453 | NP_006236.1 | |
| NM_180976.2 | NP_851307.1 | |||
| NM_180977.2 | NP_851308.1 | |||
| AB000634.1 | 3 | 288 | BAA20381.1 | |
| AB000635.1 | BAA20382.1 | |||
| AB209861.1 | 1 | 32 | BAD93098.1 | |
| AB451342.1 | 3 | 274 | ||
| AB451357.1 | 3 | 274 | ||
| AK122701.1 | 3 | 347 | ||
| AK290604.1 | ||||
| AK291797.1 | 3 | 361 | ||
| AK299696.1 | 3 | 326 | ||
| BC001095.2 | 3 | 337 | AAH01095.1 | |
| BC001175.1 | AAH01175.1 | |||
| BC010692.1 | 3 | 347 | AAH10692.1 | |
| D78360.1 | BAA11372.1 | |||
| DA388594.1 | 3 | 453 | ||
| L76702.1 | 3 | 461 | AAB69751.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3576145 | Chr.6:42728097 - 43012090 on Build GRCh38 | Gain |
|
| nsv462927 | Chr.6:42948182 - 43110718 on Build GRCh38 | Loss |
|
| nsv1017398 | Chr.6:42659705 - 43400998 on Build GRCh38 | Gain |
|
| nsv462930 | Chr.6:43002769 - 43110718 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map