Assay Details
Target Gene Details
Entrez Gene ID: | 8890 |
Gene Name: | eukaryotic translation initiation factor 2B subunit delta |
Gene Aliases: |
EIF-2B, EIF2B, EIF2Bdelta |
Location: |
Chr.2:27364352-27370457 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF2B4 | NM_001034116.1 | 4 | 473 | NP_001029288.1 |
| NM_001318965.1 | 3 | 432 | NP_001305894.1 | |
| NM_001318966.1 | 4 | 523 | NP_001305895.1 | |
| NM_001318967.1 | 4 | 466 | NP_001305896.1 | |
| NM_001318968.1 | NP_001305897.1 | |||
| NM_001318969.1 | 4 | 473 | NP_001305898.1 | |
| NM_015636.3 | 4 | 470 | NP_056451.3 | |
| NM_172195.3 | 3 | 429 | NP_751945.2 | |
| XM_006712132.1 | 4 | 520 | XP_006712195.1 | |
| XM_011533147.2 | 4 | 354 | XP_011531449.1 | |
| AB209532.1 | 2 | 1026 | BAD92769.1 | |
| AF057699.1 | 4 | 340 | AAQ13438.1 | |
| AF112207.1 | 4 | 349 | AAF17195.1 | |
| AF218009.1 | 4 | 494 | AAG17251.1 | |
| AJ011305.1 | 4 | 346 | CAB57260.1 | |
| AJ011306.1 | 3 | 429 | CAB57261.1 | |
| AK300734.1 | 4 | 520 | ||
| BC001870.1 | 4 | 331 | AAH01870.1 | |
| BC091502.1 | 4 | 346 | AAH91502.1 | |
| BG717490.1 | 3 | 411 | ||
| BI255289.1 | ||||
| JQ991007.1 | 4 | 473 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833614 | Chr.2:27263559 - 27452213 on Build GRCh38 | Gain |
|
| esv3575209 | Chr.2:27334578 - 27389381 on Build GRCh38 | Gain |
|
| nsv581244 | Chr.2:27335385 - 27480628 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map