Assay Details
Target Gene Details
Entrez Gene ID: | 7818 |
Gene Name: | death associated protein 3 |
Gene Aliases: |
DAP-3, MRP-S29, MRPS29, bMRP-10 |
Location: |
Chr.1:155688104-155739010 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DAP3 | NM_001199849.1 | 6 | 498 | NP_001186778.1 |
| NM_001199850.1 | 4 | 303 | NP_001186779.1 | |
| NM_001199851.1 | 4 | 282 | NP_001186780.1 | |
| NM_004632.3 | 5 | 402 | NP_004623.1 | |
| NM_033657.2 | 5 | 471 | NP_387506.1 | |
| XM_005245480.2 | 4 | 368 | XP_005245537.1 | |
| XM_005245481.2 | 4 | 347 | XP_005245538.1 | |
| XM_005245482.4 | 3 | 182 | XP_005245539.1 | |
| XM_011509972.2 | 5 | 401 | XP_011508274.1 | |
| XM_017002289.1 | 5 | 470 | XP_016857778.1 | |
| XM_017002290.1 | 6 | 494 | XP_016857779.1 | |
| XM_017002291.1 | 5 | 388 | XP_016857780.1 | |
| XM_017002292.1 | 5 | 367 | XP_016857781.1 | |
| XM_017002293.1 | 4 | 286 | XP_016857782.1 | |
| XM_017002294.1 | 5 | 388 | XP_016857783.1 | |
| XM_017002295.1 | 5 | 367 | XP_016857784.1 | |
| XM_017002296.1 | 4 | 347 | XP_016857785.1 | |
| XM_017002297.1 | 3 | 245 | XP_016857786.1 | |
| AK225869.1 | 6 | 498 | ||
| AK293117.1 | 5 | 403 | ||
| AK298201.1 | 4 | 290 | ||
| AK302281.1 | 4 | 303 | ||
| BC107487.1 | 5 | 402 | AAI07488.1 | |
| BC107488.1 | 5 | 471 | AAI07489.1 | |
| BT019494.1 | 4 | 314 | AAV38301.1 | |
| CR749790.1 | 5 | 482 | CAH18651.1 | |
| U18321.1 | 5 | 483 | AAA57443.1 | |
| X83544.1 | 5 | 387 | CAA58535.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33869 | Chr.1:155253492 - 155948170 on Build GRCh38 | Loss |
|
| nsv428246 | Chr.1:155588406 - 155816040 on Build GRCh38 | Gain |
|
| esv2718429 | Chr.1:155648347 - 155763436 on Build GRCh38 | Deletion |
|
| nsv3065 | Chr.1:155712977 - 155757733 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map