Assay Details
Target Gene Details
Entrez Gene ID: | 54476 |
Gene Name: | ring finger protein 216 |
Gene Aliases: |
CAHH, TRIAD3, U7I1, UBCE7IP1, ZIN |
Location: |
Chr.7:5620041-5781730 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF216 | NM_207111.3 | 17 | 3189 | NP_996994.1 |
| NM_207116.2 | 17 | 3018 | NP_996999.1 | |
| XM_005249785.2 | 19 | 3509 | XP_005249842.1 | |
| XM_011515436.1 | 15 | 2928 | XP_011513738.1 | |
| XM_017012363.1 | 17 | 2873 | XP_016867852.1 | |
| XM_017012365.1 | 16 | 2831 | XP_016867854.1 | |
| AF228527.1 | AAF36723.1 | |||
| AF513717.1 | AAP47174.1 | |||
| AF513718.1 | AAP47175.1 | |||
| AK000916.1 | BAA91422.1 | |||
| AK024787.1 | 3 | 761 | ||
| AK123635.1 | ||||
| AK292396.1 | 17 | 3198 | ||
| AK297231.1 | 8 | 1143 | ||
| AY062174.1 | AAL38043.1 | |||
| AY177396.1 | AAO60361.1 | |||
| AY177397.1 | AAO60362.1 | |||
| AY177398.1 | AAO60363.1 | |||
| BC000787.2 | 11 | 1603 | AAH00787.2 | |
| BC063825.1 | AAH63825.1 | |||
| BX537406.1 | CAD97648.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830890 | Chr.7:5425302 - 5665527 on Build GRCh38 | Loss |
|
| nsv436904 | Chr.7:5035574 - 6713282 on Build GRCh38 | Insertion |
|
| nsv1034883 | Chr.7:5612049 - 5656516 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map