Assay Details
Target Gene Details
Entrez Gene ID: | 2979 |
Gene Name: | guanylate cyclase activator 1B |
Gene Aliases: |
GCAP2, GUCA2, RP48 |
Location: |
Chr.6:42183284-42194956 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GUCA1B | NM_002098.5 | NP_002089.4 | ||
| BQ636025.1 | ||||
| BX537393.1 | CAD97635.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830649 | Chr.6:42047596 - 42231697 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map