Assay Details
Target Gene Details
Entrez Gene ID: | 25960 |
Gene Name: | adhesion G protein-coupled receptor A2 |
Gene Aliases: |
GPR124, TEM5 |
Location: |
Chr.8:37796883-37843986 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADGRA2 | NM_032777.9 | NP_116166.9 | ||
| AI346793.1 |
Target Gene Details
Entrez Gene ID: | 55290 |
Gene Name: | BRF2, RNA polymerase III transcription initiation factor 50 kDa subunit |
Gene Aliases: |
BRFU, TFIIIB50 |
Location: |
Chr.8:37843880-37849913 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BRF2 | NM_018310.3 | 4 | 1849 | NP_060780.2 |
| AA496239.1 | 1 | 159 | ||
| AF130058.1 | 1 | 929 | AAG35486.1 | |
| AF206673.2 | 4 | 1849 | AAG35669.2 | |
| AK001914.1 | 4 | 1811 | BAA91975.1 | |
| BC010648.1 | 4 | 1802 | AAH10648.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509261 | Chr.8:37767151 - 37881434 on Build GRCh38 | Insertion |
|
| nsv528598 | Chr.8:37803553 - 37859603 on Build GRCh38 | Loss |
|
| nsv951126 | Chr.8:37827983 - 37845682 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map