Assay Details
Target Gene Details
Entrez Gene ID: | 23389 |
Gene Name: | mediator complex subunit 13 like |
Gene Aliases: |
MRFACD, PROSIT240, THRAP2, TRAP240L |
Location: |
Chr.12:115958576-116277219 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 29 - Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MED13L | NM_015335.4 | NP_056150.1 | ||
| XM_011538080.2 | XP_011536382.1 | |||
| XM_011538081.2 | XP_011536383.1 | |||
| XM_011538082.2 | XP_011536384.1 | |||
| XM_017019090.1 | XP_016874579.1 | |||
| AB028948.2 | BAA82977.2 | |||
| AF515599.1 | AAQ08182.1 | |||
| AK023837.1 | BAB14697.1 | |||
| AL133033.1 | CAB61363.1 | |||
| AL137644.1 | CAB70855.1 | |||
| AY338463.1 | AAR08418.1 | |||
| BC130422.1 | ||||
| CR749332.1 | CAH18186.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3580482 | Chr.12:115964676 - 115969198 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map