Assay Details
Target Gene Details
Entrez Gene ID: | 151050 |
Gene Name: | KAT8 regulatory NSL complex subunit 1 like |
Gene Aliases: |
C2orf67, MSL1v2 |
Location: |
Chr.2:210021421-210171370 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KANSL1L | NM_001307976.1 | 14 | 3603 | NP_001294905.1 |
NM_152519.3 | 15 | 3729 | NP_689732.2 | |
XM_005246328.4 | 15 | 4011 | XP_005246385.1 | |
XM_005246329.4 | 15 | 3717 | XP_005246386.1 | |
XM_005246330.4 | 15 | 3598 | XP_005246387.1 | |
XM_005246332.4 | 14 | 3884 | XP_005246389.1 | |
XM_011510706.2 | 15 | 3636 | XP_011509008.1 | |
XM_011510707.2 | 15 | 3819 | XP_011509009.1 | |
XM_011510710.2 | 14 | 2468 | XP_011509012.1 | |
XM_017003430.1 | 15 | 3755 | XP_016858919.1 | |
XM_017003431.1 | 11 | 5837 | XP_016858920.1 | |
AA404520.1 | 1 | 286 | ||
AK074441.1 | 14 | 2498 | BAB85085.1 | |
AL133053.1 | 9 | 1721 | CAB61379.1 | |
BC126155.1 | ||||
BC143914.1 | ||||
CA393360.1 | 1 | 386 |
Target Gene Details
Entrez Gene ID: | 6120 |
Gene Name: | ribulose-5-phosphate-3-epimerase |
Gene Aliases: |
RPE2-1 |
Location: |
Chr.2:210002565-210029156 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RPE | XM_006712677.3 | 7 | 3375 | XP_006712740.1 |
AK055724.1 |

More Information
Additional Information:
For this assay, SNP(s) [rs371200259,rs73063916] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
