Assay Details
Target Gene Details
Entrez Gene ID: | 10985 |
Gene Name: | GCN1, eIF2 alpha kinase activator homolog |
Gene Aliases: |
GCN1L, GCN1L1, PRIC295 |
Location: |
Chr.12:120127210-120194710 on Build GRCh38 |
Assay Gene Location: | Within Exon 53 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GCN1 | NM_006836.1 | 53 | 7272 | NP_006827.1 |
| BC021129.1 | 2 | 144 | AAH21129.1 | |
| BC032335.1 | 2 | 99 | AAH32335.1 | |
| BC064346.1 | AAH64346.1 | |||
| BC153881.1 | 53 | 7272 | ||
| D86973.2 | 53 | 7272 | ||
| U77700.1 | 31 | 4737 | AAC51648.1 | |
| U88836.1 | 14 | 2230 | AAD00655.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519657 | Chr.12:120095892 - 120238287 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map