Assay Details
Target Gene Details
Entrez Gene ID: | 10846 |
Gene Name: | phosphodiesterase 10A |
Gene Aliases: |
ADSD2, HSPDE10A, IOLOD, LINC00473, PDE10A19 |
Location: |
Chr.6:165327287-165988121 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 30 - Exon 31 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDE10A | NM_001130690.2 | NP_001124162.1 | ||
| NM_006661.3 | NP_006652.1 | |||
| XM_006715321.3 | XP_006715384.1 | |||
| XM_011535387.2 | XP_011533689.2 | |||
| XM_011535388.2 | XP_011533690.1 | |||
| XM_011535393.2 | XP_011533695.1 | |||
| XM_017010194.1 | XP_016865683.1 | |||
| XM_017010195.1 | XP_016865684.1 | |||
| XM_017010196.1 | XP_016865685.1 | |||
| AB020593.1 | BAA78034.1 | |||
| AB026816.1 | BAA84467.1 | |||
| AF127479.1 | AAD32595.1 | |||
| AF127480.1 | AAD32596.1 | |||
| AK021956.1 | ||||
| AK297555.1 | ||||
| BC104858.1 | AAI04859.1 | |||
| BC104860.1 | AAI04861.1 | |||
| CR536567.1 | CAG38804.1 | |||
| KU232567.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5586 | Chr.6:165280959 - 165333728 on Build GRCh38 | Deletion |
|
| nsv508438 | Chr.6:165289866 - 165347160 on Build GRCh38 | Deletion |
|
| nsv605245 | Chr.6:165129197 - 165432415 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map