Assay Details
Target Gene Details
Entrez Gene ID: | 5368 |
Gene Name: | prepronociceptin |
Gene Aliases: |
N/OFQ, NOP, OFQ, PPNOC, ppN/OFQ |
Location: |
Chr.8:28316979-28343355 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PNOC | NM_006228.4 | NP_006219.1 | ||
| XM_005273532.2 | XP_005273589.1 | |||
| XM_011544559.2 | XP_011542861.1 | |||
| AK289633.1 | ||||
| AY335948.1 | AAP94601.1 | |||
| BC034758.1 | AAH34758.1 | |||
| DA403369.1 | ||||
| U48263.1 | AAC50651.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6134 | Chr.8:28294984 - 28330125 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map