Assay Details
Target Gene Details
Entrez Gene ID: | 4926 |
Gene Name: | nuclear mitotic apparatus protein 1 |
Gene Aliases: |
NMP-22, NUMA |
Location: |
Chr.11:72002864-72080693 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 30 - Exon 31 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428262 | Chr.11:71652593 - 72093009 on Build GRCh38 | Gain+Loss |
|
| nsv951037 | Chr.11:72000055 - 72016354 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map