Assay Details
Target Gene Details
Entrez Gene ID: | 4015 |
Gene Name: | lysyl oxidase |
Gene Aliases: |
- |
Location: |
Chr.5:122063195-122078501 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOX | NM_002317.6 | 1 | 881 | NP_002308.2 |
| AF039291.1 | 1 | 614 | AAD02130.1 | |
| AK297709.1 | 2 | 505 | ||
| AK298781.1 | 2 | 519 | ||
| AK311354.1 | 1 | 639 | ||
| AK312269.1 | 1 | 667 | ||
| BC074820.2 | 1 | 387 | AAH74820.1 | |
| BC074872.2 | 1 | 387 | AAH74872.1 | |
| BC089436.1 | 1 | 653 | AAH89436.1 | |
| M94054.1 | 1 | 442 | AAA59525.1 | |
| S45875.1 | 1 | 365 | AAB23549.1 | |
| S78694.1 | 1 | 595 | AAB21243.1 |
More Information
Set Membership: |
|

Genomic Map