Assay Details
Target Gene Details
Entrez Gene ID: | 57678 |
Gene Name: | glycerol-3-phosphate acyltransferase, mitochondrial |
Gene Aliases: |
GPAT, GPAT1 |
Location: |
Chr.10:112149864-112215395 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPAM | NM_001244949.1 | 22 | 5262 | NP_001231878.1 |
| NM_020918.5 | 22 | 5258 | NP_065969.3 | |
| XM_005269998.1 | 22 | 5403 | XP_005270055.1 | |
| AB046780.1 | 16 | 4550 | BAB13386.1 | |
| AK025752.1 | 1 | 1084 | ||
| AL833061.2 | 22 | 5244 | CAD89967.1 | |
| BQ694715.1 | ||||
| DA728196.1 | 1 | 339 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508608 | Chr.10:112030892 - 112158412 on Build GRCh38 | Deletion |
|
| nsv831989 | Chr.10:112112194 - 112301601 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map