Assay Details
Target Gene Details
Entrez Gene ID: | 406 |
Gene Name: | aryl hydrocarbon receptor nuclear translocator like |
Gene Aliases: |
BMAL1, BMAL1c, JAP3, MOP3, PASD3, TIC, bHLHe5 |
Location: |
Chr.11:13276613-13387268 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv553552 | Chr.11:13276972 - 13278110 on Build GRCh38 | Gain |
|
| esv24912 | Chr.11:13277442 - 13278142 on Build GRCh38 | Loss |
|
| nsv553554 | Chr.11:13277469 - 13278110 on Build GRCh38 | Gain |
|
| dgv1658n54 | Chr.11:13276938 - 13277953 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map