Assay Details
Target Gene Details
Entrez Gene ID: | 57647 |
Gene Name: | DEAH-box helicase 37 |
Gene Aliases: |
DDX37 |
Location: |
Chr.12:124946824-124989125 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DHX37 | NM_032656.3 | 20 | 2720 | NP_116045.2 |
| XM_005253590.3 | 20 | 2724 | XP_005253647.1 | |
| XM_011538598.2 | 20 | 2724 | XP_011536900.1 | |
| XM_011538600.2 | 20 | 2714 | XP_011536902.1 | |
| AB040950.1 | 17 | 2092 | BAA96041.1 | |
| BC002575.2 | 17 | 2103 | AAH02575.2 | |
| BC037964.1 | 20 | 2702 | AAH37964.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv560595 | Chr.12:124737381 - 125019207 on Build GRCh38 | Gain |
|
| nsv520994 | Chr.12:124927460 - 124961890 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map