Assay Details
Target Gene Details
Entrez Gene ID: | 164668 |
Gene Name: | apolipoprotein B mRNA editing enzyme catalytic subunit 3H |
Gene Aliases: |
A3H, ARP-10, ARP10 |
Location: |
Chr.22:39097204-39104067 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APOBEC3H | NM_001166002.2 | 5 | 827 | NP_001159474.2 |
| NM_001166003.2 | 6 | 924 | NP_001159475.2 | |
| NM_181773.4 | 5 | 830 | NP_861438.3 | |
| XM_011529990.2 | 6 | 966 | XP_011528292.1 | |
| XM_011529991.2 | 6 | 963 | XP_011528293.1 | |
| XM_011529992.2 | 6 | 940 | XP_011528294.1 | |
| BC069023.1 | 5 | 812 | AAH69023.1 | |
| BQ052182.1 | 5 | 769 | ||
| BQ054330.1 | 6 | 870 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834197 | Chr.22:39077294 - 39214999 on Build GRCh38 | Gain |
|
| nsv510807 | Chr.22:39075404 - 39147379 on Build GRCh38 | Deletion |
|
| nsv834196 | Chr.22:38996564 - 39214999 on Build GRCh38 | Loss |
|
| nsv1066354 | Chr.22:39070438 - 39130755 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map