Assay Details
Target Gene Details
Entrez Gene ID: | 387885 |
Gene Name: | cilia and flagella associated protein 73 |
Gene Aliases: |
CCDC42B, MIA2 |
Location: |
Chr.12:113149162-113159276 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CFAP73 | NM_001144872.1 | NP_001138344.1 |
Target Gene Details
Entrez Gene ID: | 79039 |
Gene Name: | DEAD-box helicase 54 |
Gene Aliases: |
DP97 |
Location: |
Chr.12:113157173-113185479 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX54 | NM_001111322.1 | 20 | 2958 | NP_001104792.1 |
| NM_024072.3 | 20 | 2955 | NP_076977.3 | |
| AF478457.1 | 20 | 2958 | AAL85336.1 | |
| AY148094.1 | 20 | 2934 | AAN59978.1 | |
| BC005848.2 | 20 | 2876 | AAH05848.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519540 | Chr.12:113042705 - 113459599 on Build GRCh38 | Gain+Loss |
|
| esv3892265 | Chr.12:113099321 - 113527932 on Build GRCh38 | Gain |
|
| dgv2884n54 | Chr.12:113115603 - 113166364 on Build GRCh38 | Loss |
|
| esv3630792 | Chr.12:112988703 - 113838810 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map