Assay Details
Target Gene Details
Entrez Gene ID: | 8613 |
Gene Name: | phospholipid phosphatase 3 |
Gene Aliases: |
Dri42, LPP3, PAP2B, PPAP2B, VCIP |
Location: |
Chr.1:56494747-56579584 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLPP3 | NM_003713.4 | 6 | 1467 | NP_003704.3 |
| AB000889.1 | 6 | 905 | BAA22594.1 | |
| AF017786.1 | 7 | 1273 | AAC63433.1 | |
| AF043329.1 | 6 | 1192 | AAD02271.1 | |
| AF480883.1 | 7 | 1279 | AAO84481.1 | |
| AK123105.1 | 2 | 1885 | ||
| AK312439.1 | 6 | 1425 | ||
| BC009196.2 | 6 | 1270 | AAH09196.1 | |
| BT019589.1 | 6 | 899 | AAV38396.1 | |
| U79294.1 | 7 | 1272 | AAB50222.1 |
More Information
Set Membership: |
|

Genomic Map