Assay Details
Target Gene Details
Entrez Gene ID: | 8841 |
Gene Name: | histone deacetylase 3 |
Gene Aliases: |
HD3, RPD3, RPD3-2 |
Location: |
Chr.5:141620876-141636860 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HDAC3 | NM_003883.3 | 15 | 1363 | NP_003874.2 |
| XM_011537697.2 | 14 | 1215 | XP_011535999.1 | |
| AF005482.1 | 13 | 1379 | AAB87752.1 | |
| AF039703.1 | 15 | 1319 | AAC98927.1 | |
| BC000614.1 | 15 | 1352 | AAH00614.1 | |
| U66914.1 | 15 | 1308 | AAC52038.1 | |
| U75696.1 | 14 | 1346 | AAB88240.1 | |
| U75697.1 | 15 | 1352 | AAB88241.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5033 | Chr.5:141599131 - 141630987 on Build GRCh38 | Insertion |
|
| nsv516495 | Chr.5:141576210 - 141624764 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map