Assay Details
Target Gene Details
Entrez Gene ID: | 55830 |
Gene Name: | glycosyltransferase 8 domain containing 1 |
Gene Aliases: |
AD-017, MSTP139 |
Location: |
Chr.3:52694484-52706083 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLT8D1 | NM_001010983.2 | NP_001010983.1 | ||
| NM_001278280.1 | NP_001265209.1 | |||
| NM_001278281.1 | NP_001265210.1 | |||
| NM_018446.3 | NP_060916.1 | |||
| NM_152932.2 | 1 | 112 | NP_690909.1 | |
| XM_017006857.1 | 3 | 942 | XP_016862346.1 | |
| XM_017006858.1 | 2 | 713 | XP_016862347.1 | |
| XM_017006859.1 | XP_016862348.1 | |||
| AF157318.1 | AAF67484.1 | |||
| AF175227.1 | AAQ13652.1 | |||
| AF176777.1 | AAQ13655.1 | |||
| AK027517.1 | BAB55170.1 | |||
| AK311674.1 | 3 | 675 | ||
| AY358579.1 | AAQ88942.1 | |||
| BC119670.1 | ||||
| BC119671.1 | ||||
| BF725274.1 | ||||
| BG700867.1 | ||||
| CR749821.1 | CAH18681.1 |
More Information
Set Membership: |
|

Genomic Map