Assay Details
Target Gene Details
Entrez Gene ID: | 5175 |
Gene Name: | platelet and endothelial cell adhesion molecule 1 |
Gene Aliases: |
CD31, CD31/EndoCAM, GPIIA', PECA1, PECAM-1, endoCAM |
Location: |
Chr.17:64319415-64413844 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PECAM1 | NM_000442.4 | NP_000433.4 | ||
| XM_005276880.1 | XP_005276937.1 | |||
| XM_005276881.1 | XP_005276938.1 | |||
| XM_005276882.1 | XP_005276939.1 | |||
| XM_005276883.2 | XP_005276940.1 | |||
| XM_011524889.2 | XP_011523191.1 | |||
| XM_011524890.1 | XP_011523192.1 | |||
| XM_017024738.1 | XP_016880227.1 | |||
| XM_017024739.1 | XP_016880228.1 | |||
| XM_017024740.1 | XP_016880229.1 | |||
| XM_017024741.1 | XP_016880230.1 | |||
| AK290692.1 | ||||
| AK303959.1 | ||||
| AK304166.1 | ||||
| BC022512.1 | AAH22512.1 | |||
| BC051822.1 | AAH51822.1 | |||
| JQ287500.1 | ||||
| M28526.1 | AAA36429.1 | |||
| M37780.1 | AAA36186.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1066968 | Chr.17:64215762 - 64332226 on Build GRCh38 | Gain |
|
| nsv833513 | Chr.17:64235366 - 64396432 on Build GRCh38 | Loss |
|
| nsv524852 | Chr.17:64215039 - 64329779 on Build GRCh38 | Gain |
|
| nsv833512 | Chr.17:64210750 - 64396432 on Build GRCh38 | Gain+Loss |
|
| nsv509666 | Chr.17:64254455 - 64396455 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map